2-Chloro-4-iodophenol 97%

Supplier: Ambeed
Warning

A103766-10G A103766-1G A103766-5G A103766-250MG
103796-606EA 30 USD
103796-606 103822-316 76869-278 77283-158 77283-160 77882-888
2-Chloro-4-iodophenol 97%
2-Chloro-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄ClIO
Storage Temperature: Refrigerator
MDL Number: MFCD11110536
CAS Number: 116130-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR