2-Chloro-4-iodo-5-methylpyridine 97%

Supplier: Ambeed
Warning

A143938-100MG A143938-10G A143938-250MG A143938-5G A143938-1G
103803-354EA 41 USD
103803-354 76871-342 76871-344 76871-346 76871-348 77316-662
2-Chloro-4-iodo-5-methylpyridine 97%
2-Chloro-4-iodo-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅ClIN
Boiling Pt: 277.4±35.0 °C (760 mm Hg)
Flash Pt: 121.6 °C
Storage Temperature: Freezer
MDL Number: MFCD16610529
CAS Number: 1197957-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR