2-Chloro-4-iodo-3-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-iodo-3-picoline

A249992-10G A249992-250MG A249992-1G A249992-25G
103811-904EA 23 USD
103811-904 103851-744 76647-748 76647-750 77091-094 77304-510
2-Chloro-4-iodo-3-methylpyridine 97%
2-Chloro-4-iodo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅ClIN
MW: 253.47
Boiling Pt: 274.9 °C (760 mm Hg)
Melting Pt: 101…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095293
CAS Number: 153034-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR