2-Chloro-4-hydroxypyridine 98%

Supplier: Ambeed
Warning

A167170-25G A167170-100G A167170-10G A167170-5G
103844-402EA 12 USD
103844-402 77081-402 77286-638 77286-640 77300-876
2-Chloro-4-hydroxypyridine 98%
2-Chloro-4-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄ClNO
Boiling Pt: 225.1 °C (760 mm Hg)
Melting Pt: 169…172 °C
Storage Temperature: Ambient
MDL Number: MFCD01646108
CAS Number: 17368-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR