2-Chloro-4-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A472234-100G A472234-10G A472234-250MG A472234-1G A472234-25G A472234-5G
103828-042EA 11 USD
103828-042 103839-116 76774-288 76774-290 76774-292 77307-492 77307-494 77307-496
2-Chloro-4-hydroxybenzaldehyde 97%
2-Chloro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 272.5 °C (760 mm Hg)
Melting Pt: 145…147 °C
Flash Pt: 118.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00052184
CAS Number: 56962-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR