2-Chloro-4-fluoropyridine 97%

Supplier: Ambeed
Danger

A632035-1G A632035-25G A632035-5G A632035-10G
103840-278EA 17 USD
103840-278 76844-446 76844-448 76844-450 76992-262
2-Chloro-4-fluoropyridine 97%
2-Chloro-4-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClFN
Boiling Pt: 157.9 °C (760 mm Hg)
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039345
CAS Number: 34941-91-8
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR