2-Chloro-4-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A258145-1G A258145-25G A258145-100G A258145-10G A258145-5G
103812-362EA 9 USD
103812-362 103852-236 77091-762 77091-764 77098-780 77098-782 77304-728
2-Chloro-4-fluorophenylboronic acid 98%
2-Chloro-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClFO₂
Boiling Pt: 290.9 °C (760 mm Hg)
Melting Pt: 220…223 °C
Flash Pt: 129.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02684295
CAS Number: 313545-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR