2-Chloro-4-fluorophenol 98%

Supplier: Ambeed
Warning

A223158-100G A223158-10G A223158-500G A223158-25G A223158-5G
103849-904EA 9 USD
103849-904 76999-988 76999-990 76999-992 77088-520 77303-646
2-Chloro-4-fluorophenol 98%
2-Chloro-4-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 227.6 °C (760 mm Hg)
Melting Pt: 23 °C
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00002168
CAS Number: 1996-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR