1-Bromo-2-chloro-4-fluorobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-3-chloro-1-fluorobenzene, 2-Chloro-4-fluorobromobenzene, 4-Bromo-3-chlorofluorobenzene
A320087-25G
A320087-5G
A320087-100G
A320087-10G
A320087-500G
A320087-1G
103825-228EA
5
USD
103825-228
103836-160
76839-024
76839-026
77154-490
77154-492
77154-494
77306-208
1-Bromo-2-chloro-4-fluorobenzene 98%
1-Bromo-2-chloro-4-fluorobenzene
Precaución: for Research Use Only
Formula:
C₆H₃BrClF MW: 209.44 g/mol Boiling Pt: 196.3 °C (760 mm Hg) Flash Pt: 72.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142587 CAS Number: 110407-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...