2-Chloro-4-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A240792-100G A240792-500G A240792-1000G A240792-25G
103809-708EA 343 USD
103809-708 103851-168 76771-402 76771-404 77090-320 77090-322
2-Chloro-4-fluorobenzaldehyde 98%
2-Chloro-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO
Boiling Pt: 118…120 °C (760 mm Hg)
Melting Pt: 60…63 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042527
CAS Number: 84194-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR