2-Chloro-4-fluoro-5-nitrobenzoic acid 96%

Supplier: Ambeed
Warning

A137092-100G A137092-10G A137092-1G A137092-25G A137092-5G
103802-400EA 5 USD
103802-400 77258-696 77258-698 77315-348 77315-350 77315-352
2-Chloro-4-fluoro-5-nitrobenzoic acid 96%
2-Chloro-4-fluoro-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClFNO
Boiling Pt: 364.2 °C (760 mm Hg)
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD04115715
CAS Number: 114776-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR