2-Chloro-4-fluoro-5-methylaniline 97%
Supplier: Ambeed
|
Warning
|
A222319-250MG
A222319-1G
A222319-5G
A222319-10G
103818-972EA
6
USD
103818-972
103849-852
76770-868
77088-458
77303-622
77877-690
2-Chloro-4-fluoro-5-methylaniline 97%
2-Chloro-4-fluoro-5-methylaniline
Precaución: for Research Use Only
Formula:
C₇H₇ClFN Melting Pt: 70…73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00055369 CAS Number: 124185-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...