2-Chloro-4-ethoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-ethoxybenzeneboronic acid

A229199-1G A229199-250MG
103819-452EA 21 USD
103819-452 103850-354 77089-178 77193-086
2-Chloro-4-ethoxyphenylboronic acid 97%
2-Chloro-4-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₃
MW: 200.43
Boiling Pt: 352.8 °C (760 mm Hg)
Flash Pt: 167.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02684318
CAS Number: 313545-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR