2-Chloro-4-cyanobenzoic acid 97%

Supplier: Ambeed
Warning

A153399-100MG A153399-250MG
103804-508EA 45 USD
103804-508 103842-680 77010-910 77010-912
2-Chloro-4-cyanobenzoic acid 97%
2-Chloro-4-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂
MW: 181.58 g/mol
Boiling Pt: 338.1±27.0 °C (760 mm Hg)
Melting Pt: 170…171 °C
Flash Pt: 158.3 °C
Storage Temperature: Ambient
MDL Number: MFCD11036377
CAS Number: 117738-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR