2-Chloro-4,6-diphenylpyrimidine 97%

Supplier: Ambeed
Warning

A112665-250MG A112665-1G
103798-066EA 6 USD
103798-066 103823-904 77257-744 77310-718
2-Chloro-4,6-diphenylpyrimidine 97%
2-Chloro-4,6-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₆H₁₁ClN₂
MW: 266.73 g/mol
Boiling Pt: 469.1 °C (760 mm Hg)
Flash Pt: 269.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00030776
CAS Number: 2915-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR