2-Chloro-4,6-diphenyl-1,3,5-triazine 98%
Supplier: Ambeed
|
Warning
|
A413484-5G
A413484-100G
A413484-10G
A413484-500G
A413484-1G
A413484-25G
103827-260EA
6
USD
103827-260
103838-306
76841-862
77033-768
77033-770
77033-772
77307-164
77307-166
2-Chloro-4,6-diphenyl-1,3,5-triazine 98%
2-Chloro-4,6-diphenyl-1,3,5-triazine
Precaución: for Research Use Only
Formula:
C₁₅H₁₀ClN₃ MW: 267.72 g/mol Boiling Pt: 485.1 °C (760 mm Hg) Melting Pt: 138…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD05738885 CAS Number: 3842-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...