2-Chloro-4,6-dimethylpyridine 97%

Supplier: Ambeed
Warning

A171226-25G A171226-5G A171226-10G A171226-1G
103806-604EA 11 USD
103806-604 103844-880 77082-014 77082-016 77096-528 77301-108
2-Chloro-4,6-dimethylpyridine 97%
2-Chloro-4,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.60 g/mol
Boiling Pt: 208.1 °C (760 mm Hg)
Flash Pt: 99.11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08277279
CAS Number: 30838-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR