2-Chloro-4,6-dimethyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

A157998-100MG A157998-250MG A157998-1G
103805-080EA 320 USD
103805-080 77011-070 77011-072 77300-308
2-Chloro-4,6-dimethyl-3-nitropyridine 98%
2-Chloro-4,6-dimethyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.596 g/mol
Melting Pt: 187 °C
Storage Temperature: Ambient
MDL Number: MFCD18261551
CAS Number: 89793-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR