2-Chloro-4,6-dimethoxypyridine 97%

Supplier: Ambeed
Warning

A248807-250MG A248807-1G
103811-840EA 114 USD
103811-840 103851-674 76874-566 77304-464
2-Chloro-4,6-dimethoxypyridine 97%
2-Chloro-4,6-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11113389
CAS Number: 108279-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR