2-Chloro-4,5-difluorobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A133197-10G
A133197-1G
A133197-5G
103801-842EA
24
USD
103801-842
76767-324
76767-326
77314-466
77314-468
2-Chloro-4,5-difluorobenzaldehyde 97%
2-Chloro-4,5-difluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃ClF₂O Flash Pt: 81.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07782050 CAS Number: 165047-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...