2-Chloro-4-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A238174-1G A238174-25G A238174-5G A238174-100G A238174-10G A238174-500G
103811-204EA 9 USD
103811-204 103850-988 77090-096 77090-098 77090-100 77288-654 77288-656 77615-966
2-Chloro-4-(trifluoromethyl)pyridine 98%
2-Chloro-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 146…147 °C (760 mm Hg)
Melting Pt: …19 °C
Storage Temperature: Ambient
MDL Number: MFCD00042224
CAS Number: 81565-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR