2-Chloro-4-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-cyanobenzotrifluoride

A247204-25G A247204-5G A247204-100G A247204-10G
103851-590EA 14 USD
103851-590 77090-912 77090-914 77193-532 77193-534
2-Chloro-4-(trifluoromethyl)benzonitrile 98%
2-Chloro-4-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃ClF₃N
Boiling Pt: 192…193 °C (760 mm Hg)
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00084939
CAS Number: 1813-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR