2-Chloro-4-(tert-pentyl)phenol 96%

Supplier: Ambeed
Warning

A195685-1G A195685-100MG A195685-250MG A195685-25G
103816-772EA 35 USD
103816-772 103847-384 77085-336 77260-654 77260-656 77302-476
2-Chloro-4-(tert-pentyl)phenol 96%
2-Chloro-4-(tert-pentyl)phenol

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClO
MW: 198.69 g/mol
Boiling Pt: 257.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00019993
CAS Number: 5323-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR