2-Chloro-4-(methylthio)pyrimidine 98%

Supplier: Ambeed
Warning

A160325-100MG A160325-10G A160325-250MG A160325-1G A160325-5G
103805-364EA 14 USD
103805-364 103843-592 77110-634 77110-636 77110-638 77319-516 77319-518
2-Chloro-4-(methylthio)pyrimidine 98%
2-Chloro-4-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂S
MW: 160.6300 g/mol
Boiling Pt: 308.3 °C (760 mm Hg)
Melting Pt: 67…68 °C
Flash Pt: 140.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09265490
CAS Number: 49844-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR