2-Chloro-4-(methylsulfonyl)pyrimidine 97%

Supplier: Ambeed
Warning

A140748-100MG A140748-250MG
103802-898EA 87 USD
103802-898 76782-904 77600-046
2-Chloro-4-(methylsulfonyl)pyrimidine 97%
2-Chloro-4-(methylsulfonyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂S
MW: 192.62 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16659665
CAS Number: 1233026-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR