2-Chloro-4-(methoxycarbonyl)benzoic acid 95%

Supplier: Ambeed
Warning

A138428-250MG A138428-1G
103802-612EA 79 USD
103802-612 76811-640 77298-990
2-Chloro-4-(methoxycarbonyl)benzoic acid 95%
2-Chloro-4-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇ClO₄
MW: 214.6 g/mol
Boiling Pt: 360.3±27.0 °C (760 mm Hg)
Flash Pt: 171.7 °C
Storage Temperature: Ambient
MDL Number: MFCD21603679
CAS Number: 431888-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR