2-Chloro-4-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

A195076-5G A195076-100MG A195076-10G A195076-250MG A195076-1G A195076-25G A195076-100G
103816-722EA 14 USD
103816-722 103847-336 77085-286 77164-390 77164-392 77164-394 77302-442 77302-444 77892-260
2-Chloro-4-pyridineboronic acid pinacol ester 97%
2-Chloro-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₂
Boiling Pt: 338.2 °C (760 mm Hg)
Melting Pt: 64.7…64.8 °C
Flash Pt: 158.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04039870
CAS Number: 458532-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR