(2-Chloro-3-pyridinyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-pyridinemethanol, (2-chloropyridin-3-yl)methanol, 2-Chloro-3-pyridinylmethanol

A173260-100G A173260-10G A173260-25G A173260-5G A173260-1G
103806-808EA 6 USD
103806-808 103845-102 76974-130 76974-132 76974-134 77082-350 77301-252
(2-Chloro-3-pyridinyl)methanol 97%
(2-Chloro-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 276.7 °C (760 mm Hg)
Melting Pt: 54 °C
Flash Pt: 121.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01935961
CAS Number: 42330-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR