2-Chloro-3-nitropyridin-4-ol 98%

Supplier: Ambeed
Warning

A219777-1G A219777-100MG A219777-250MG A219777-5G
103818-816EA 475 USD
103818-816 77088-260 77112-696 77112-698 77112-700
2-Chloro-3-nitropyridin-4-ol 98%
2-Chloro-3-nitropyridin-4-ol

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MW: 174.5400 g/mol
Boiling Pt: 408.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07778466
CAS Number: 629655-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR