2-Chloro-3-nitrophenol 97%

Supplier: Ambeed
Danger

A147113-10G A147113-250MG A147113-1G
103803-742EA 12 USD
103803-742 103841-838 77259-086 77259-088 77317-198
2-Chloro-3-nitrophenol 97%
2-Chloro-3-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MW: 173.5500 g/mol
Boiling Pt: 279.6 °C (760 mm Hg)
Flash Pt: 122.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11035882
CAS Number: 603-84-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR