2-Chloro-3-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A279311-10G A279311-250MG A279311-1G A279311-25G A279311-5G
103813-464EA 32 USD
103813-464 103853-408 77014-580 77014-582 77305-328 77305-330 77305-332
2-Chloro-3-nitrobenzonitrile 98%
2-Chloro-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClN₂O₂
MW: 182.5600 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD11044838
CAS Number: 34662-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR