2-Chloro-3-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Mesalazine EP Impurity Q

A297597-5G A297597-100G A297597-10G A297597-250MG A297597-1G A297597-25G A297597-500G
103814-144EA 6 USD
103814-144 103835-286 76837-972 77141-464 77141-466 77141-468 77305-708 77305-710 77685-851
2-Chloro-3-nitrobenzoic acid 97%
2-Chloro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 359.9 °C (760 mm Hg)
Melting Pt: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00007069
CAS Number: 3970-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR