2-Chloro-3-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A260752-1G A260752-5G A260752-100MG A260752-250MG
103812-502EA 21 USD
103812-502 103852-388 77091-974 77091-976 77140-706 77140-708
2-Chloro-3-nitrobenzaldehyde 95%
2-Chloro-3-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClNO₃
MW: 185.5700 g/mol
Boiling Pt: 293.5 °C (760 mm Hg)
Flash Pt: 131.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08236804
CAS Number: 58755-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR