2-Chloro-3-nitro-6-phenylpyridine 97%

Supplier: Ambeed
Warning

A110696-250MG A110696-1G A110696-5G
103797-706EA 50 USD
103797-706 103823-514 77008-896 77284-026 77284-028
2-Chloro-3-nitro-6-phenylpyridine 97%
2-Chloro-3-nitro-6-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₇ClN₂O₂
Boiling Pt: 376.1±37.0 °C (760 mm Hg)
Flash Pt: 181.3 °C
Storage Temperature: Ambient
MDL Number: MFCD22380358
CAS Number: 187242-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR