2-Chloro-3-nitro-6-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A733006-100MG A733006-10G A733006-250MG A733006-1G A733006-25G A733006-5G
103840-792EA 309 USD
103840-792 76791-216 76791-218 76791-220 77308-226 77308-228 77308-230
2-Chloro-3-nitro-6-(trifluoromethyl)pyridine 95%
2-Chloro-3-nitro-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₃N₂O₂
MW: 226.5405 g/mol
Boiling Pt: 242.4 °C (760 mm Hg)
Flash Pt: 100.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09864674
CAS Number: 117519-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR