2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A113782-10G A113782-25G A113782-1G A113782-5G
103798-292EA 7 USD
103798-292 103824-136 76781-256 76781-258 77310-952 77310-954
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 97%
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₃N₂O₂
MW: 226.54 g/mol
Boiling Pt: 242.3 °C (760 mm Hg)
Flash Pt: 100.3 °C
Storage Temperature: Ambient
MDL Number: MFCD06255171
CAS Number: 72587-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR