2-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxy-4-chloro-5-nitrobenzotrifluoride, 6-Chloro-5-nitro-α,α,α-trifluoro-m-toluic acid

A171301-250MG A171301-1G A171301-25G A171301-5G
103806-620EA 33 USD
103806-620 103844-894 76813-218 77082-038 77082-040 77082-042
2-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid 97%
2-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃ClF₃NO₄
MW: 269.562 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847295
CAS Number: 22227-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR