2-Chloro-3-methylquinoxaline 97%
Supplier: Ambeed
|
Warning
|
A273909-1G
A273909-5G
A273909-100MG
A273909-250MG
103813-164EA
48
USD
103813-164
103853-090
77092-902
77092-904
77099-048
77099-050
2-Chloro-3-methylquinoxaline 97%
2-Chloro-3-methylquinoxaline
Precaución: for Research Use Only
Formula:
C₉H₇ClN₂ Boiling Pt: 259.9 °C (760 mm Hg) Melting Pt: 70…73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00023913 CAS Number: 32601-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...