2-Chloro-m-anisidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-3-methoxyaniline
A194551-1G
A194551-25G
A194551-5G
A194551-10G
A194551-250MG
103816-664EA
8
USD
103816-664
103847-274
77085-194
77085-196
77085-198
77151-976
77151-978
2-Chloro-m-anisidine 98%
2-Chloro-m-anisidine
Precaución: for Research Use Only
Formula:
C₇H₈ClNO MW: 157.6000 g/mol Boiling Pt: 252 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08691737 CAS Number: 113206-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...