2-Chloro-3-methoxy-6-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A125571-1G
A125571-5G
103800-696EA
141
USD
103800-696
77298-064
77298-066
2-Chloro-3-methoxy-6-methylpyridine 98%
2-Chloro-3-methoxy-6-methylpyridine
Precaución: for Research Use Only
Formula:
C₇H₈ClNO MW: 157.598 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09260845 CAS Number: 204378-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...