2-Chloro-3-iodopyridine 98%

Supplier: Ambeed
Warning

A267069-25G A267069-5G A267069-100G A267069-10G A267069-500G
103812-832EA 7 USD
103812-832 103852-734 77092-452 77092-454 77289-380 77289-382 77289-384
2-Chloro-3-iodopyridine 98%
2-Chloro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃ClIN
MW: 239.44 g/mol
Melting Pt: 90…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00661298
CAS Number: 78607-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR