2-Chloro-3-hydroxybenzonitrile 97%

Supplier: Ambeed
Warning

A122035-100MG A122035-250MG A122035-1G
103800-170EA 46 USD
103800-170 76822-496 76822-498 77312-296
2-Chloro-3-hydroxybenzonitrile 97%
2-Chloro-3-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClNO
MW: 153.566 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848763
CAS Number: 51786-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR