2-Chloro-3-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A238886-1G A238886-5G A238886-100G A238886-10G A238886-250MG A238886-25G
103811-252EA 13 USD
103811-252 103851-048 77090-184 77090-186 77261-746 77261-748 77261-750 77304-124
2-Chloro-3-hydroxybenzaldehyde 97%
2-Chloro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Melting Pt: 139…143 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD06798118
CAS Number: 56962-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR