2-Chloro-3-hydrazinopyrazine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Chloro-3-hydrazinylpyrazine
A562796-10G
A562796-250MG
A562796-1G
103828-756EA
22
USD
103828-756
103839-890
76819-156
76819-158
76843-992
2-Chloro-3-hydrazinopyrazine 98%
2-Chloro-3-hydrazinopyrazine
Precaución: for Research Use Only
Formula:
C₄H₈Cl₂N₄ MW: 144.5600 g/mol Boiling Pt: 299.5 °C (760 mm Hg) Melting Pt: 154…159 °C Flash Pt: 134.9 °C Storage Temperature: Freezer |
MDL Number:
MFCD08272804 CAS Number: 63286-28-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...