2-Chloro-3-fluoro-6-nitroaniline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-chloro-4-fluoronitrobenzene

A119787-10G A119787-25G A119787-1G A119787-5G
103799-838EA 10 USD
103799-838 77026-312 77026-314 77311-940 77311-942
2-Chloro-3-fluoro-6-nitroaniline 98%
2-Chloro-3-fluoro-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄ClFN₂O₂
MW: 190.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09763600
CAS Number: 948014-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR