2-Chloro-3-fluoro-6-methylpyridine 98%

Supplier: Ambeed
Warning

A333161-1G A333161-10G A333161-250MG
103825-626EA 34 USD
103825-626 103836-590 76839-598 76978-298 76978-300
2-Chloro-3-fluoro-6-methylpyridine 98%
2-Chloro-3-fluoro-6-methylpyridine

Precaución: For research use only

Formula: C₆H₅ClFN
Storage Temperature: Refrigerator
MDL Number: MFCD03095216
CAS Number: 374633-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR