2-Chloro-3-fluoro-4-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-fluoro-4-picoline

A385961-1G A385961-25G A385961-5G
103826-788EA 20 USD
103826-788 103837-828 76841-172 76841-174 76841-176
2-Chloro-3-fluoro-4-methylpyridine 97%
2-Chloro-3-fluoro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFN
Boiling Pt: 186.4 °C (760 mm Hg)
Flash Pt: 66.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112510
CAS Number: 881891-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR