2-Chloro-3-fluoro-4-iodopyridine 95%
Supplier: Ambeed
|
Warning
|
A123850-250MG
A123850-25G
A123850-1G
A123850-5G
A123850-10G
103800-454EA
15
USD
103800-454
76995-960
76995-962
77312-572
77312-574
77670-756
2-Chloro-3-fluoro-4-iodopyridine 95%
2-Chloro-3-fluoro-4-iodopyridine
Precaución: for Research Use Only
Formula:
C₅H₂ClFIN MW: 257.43 g/mol Boiling Pt: 246.1 °C (760 mm Hg) Melting Pt: 104 °C Flash Pt: 102.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972389 CAS Number: 148639-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...