2-Chloro-3-fluoro-4-hydroxypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-fluoropyridin-4-ol

A153288-250MG A153288-1G A153288-5G
103804-492EA 25 USD
103804-492 103842-664 76823-888 77318-316 77318-318
2-Chloro-3-fluoro-4-hydroxypyridine 95%
2-Chloro-3-fluoro-4-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃ClFNO
Boiling Pt: 351.2±37.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD13189885
CAS Number: 1184172-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR