1-Chloro-2,3-difluoro-6-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3,4-difluoronitrobenzene

A109074-1MG A109074-250MG A109074-1G
103797-440EA 18 USD
103797-440 103823-228 77283-802 77283-804 77310-052
1-Chloro-2,3-difluoro-6-nitrobenzene 98%
1-Chloro-2,3-difluoro-6-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MW: 193.5354 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06658262
CAS Number: 169468-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR